Invitrogen
Manufacturer Code:PA567626
Catalog # PIPA567626
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human YY2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: transcription factor yin yang 2; Transcription factor YY2; Yin and yang 2; YY-2; YY2 transcription factor ZNF631; Zinc finger protein 631
Gene Aliases: YY2; ZNF631
UniProt ID: (Human) O15391, (Mouse) Q3TTC2
Entrez Gene ID: (Human) 404281, (Mouse) 100073351
Molecular Function: transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.