Invitrogen
Manufacturer Code:PA529138
Catalog # PIPA529138
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 91 and 384 of Human YY1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DELTA INO80S NF-E1 UCRBP YIN-YANG-1 INO80 complex subunit S YY-1 Yin and Yang 1 protein delta transcription factor transcriptional repressor protein YY1; Delta transcription factor; INO80 complex subunit S; NF-E1; Transcriptional repressor protein YY1; Yin and yang 1; Yin and Yang 1 protein; YY-1
Gene Aliases: DELTA; INO80S; NF-E1; UCRBP; YIN-YANG-1; YY1
UniProt ID: (Human) P25490
Entrez Gene ID: (Human) 7528
Molecular Function:
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.