Invitrogen
Manufacturer Code:PA576378
Catalog # PIPA576378
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human YTHDF3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DF3; YTH domain family 3; YTH domain family protein 3; YTH domain family, member 3; YTH domain-containing family protein 3; YTH N(6)-methyladenosine RNA binding protein 3
Gene Aliases: 9130022A11Rik; YTHDF3
UniProt ID: (Human) Q7Z739, (Mouse) Q8BYK6
Entrez Gene ID: (Human) 253943, (Mouse) 229096, (Rat) 361920
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.