Invitrogen
Manufacturer Code:PA570853
Catalog # PIPA570853
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of human YTHDF2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9430020E02Rik; CLL-associated antigen KW-14; DF2; HGRG8; high glucose-regulated protein 8; High-glucose-regulated protein 8; Renal carcinoma antigen NY-REN-2; YTH domain family 2; YTH domain family protein 2; YTH domain family, member 2; YTH domain-containing family protein 2; YTH N(6)-methyladenosine RNA binding protein 2
Gene Aliases: 9430020E02Rik; CAHL; HGRG8; NY-REN-2; YTHDF2
UniProt ID: (Bovine) Q0VCZ3, (Human) B4E1G7, (Mouse) Q91YT7
Entrez Gene ID: (Bovine) 541050, (Horse) 100070667, (Dog) 478161, (Rabbit) 100351518, (Human) 51441, (Mouse) 213541, (Rat) 313053
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.