Proteintech
Manufacturer Code:247441AP150UL
Catalog # 50-556-390
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | YTHDF2 Fusion Protein Ag5922 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9430020E02Rik; CLL associated antigen KW 14 HGRG8 NY REN 2 YTH domain family protein 2 YTH domain family member 2 YTHDF2; CLL-associated antigen KW-14; DF2; High-glucose-regulated protein 8; Renal carcinoma antigen NY-REN-2; YTH domain family, member 2; YTH domain-containing family protein 2; YTH N(6)-methyladenosine RNA binding protein 2
Gene Aliases: CAHL; HGRG8; NY-REN-2; YTHDF2
UniProt ID: (Human) B4E1G7
Entrez Gene ID: (Human) 51441
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.