Abnova
Manufacturer Code:H00051441K
Catalog # 89-207-344
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human YTHDF2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human YTHDF2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: 9430020E02Rik; CLL-associated antigen KW-14; DF2; HGRG8 NY-REN-2; High-glucose-regulated protein 8; Renal carcinoma antigen NY-REN-2; YTH domain family, member 2; YTH domain-containing family protein 2; YTH N(6)-methyladenosine RNA binding protein 2
Gene Aliases: CAHL; HGRG8; NY-REN-2; YTHDF2
UniProt ID: (Human) B4E1G7
Entrez Gene ID: (Human) 51441
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.