Abnova
Manufacturer Code:PAB22400
Catalog # 89-124-108
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human YRDC. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant YRDC.
For Research Use Only
Protein Aliases: dopamine receptor interacting protein 3; Dopamine receptor-interacting protein 3; DRIP3; FLJ23476; FLJ26165; hIRIP; IRIP; ischemia/reperfusion inducible protein; Ischemia/reperfusion-inducible protein homolog; RP11-109P14.4; SUA5; yrdC domain containing; YrdC domain-containing protein, mitochondrial; yrdC N(6)-threonylcarbamoyltransferase domain containing
Gene Aliases: DRIP3; IRIP; SUA5; YRDC
UniProt ID: (Human) Q86U90
Entrez Gene ID: (Human) 79693
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.