Proteintech
Manufacturer Code:262951AP150UL
Catalog # 50-173-5296
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | YRDC Fusion Protein Ag23633 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dopamine receptor interacting protein 3; Dopamine receptor-interacting protein 3; DRIP3 hIRIP IRIP RP11 109P14.4 SUA5 YRDC; hIRIP; ischemia/reperfusion inducible protein; Ischemia/reperfusion-inducible protein homolog; yrdC domain containing; YrdC domain-containing protein, mitochondrial; yrdC N(6)-threonylcarbamoyltransferase domain containing
Gene Aliases: DRIP3; IRIP; SUA5; YRDC
UniProt ID: (Human) Q86U90
Entrez Gene ID: (Human) 79693
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.