Invitrogen
Manufacturer Code:PA570852
Catalog # PIPA570852
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of mouse YOD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DUBA-8; HIN-7; HIV-1-induced protease 7; HIV-induced protein-7-like protease; hsHIN7; OTU domain containing 2; OTU domain-containing protein 2; Ubiquitin thioesterase OTU1; YOD1 DUBA8 HIN7 OTUD2 Ubiquitin thioesterase OTU1 DUBA-8 HIV-1-induced protease 7 OTU domain-containing protein 2; YOD1 OTU deubiquinating enzyme 1 homolog; YOD1 OTU deubiquinating enzyme 1 homolog ( yeast); YOD1 OTU deubiquitinating enzyme 1 homologue
Gene Aliases: 9930028C20Rik; DUBA8; fe01a10; HIN7; Hshin7; OTUD2; PRO0907; wu:fe01a10; YOD1; zgc:112182
UniProt ID: (Human) Q5VVQ6, (Bovine) Q05B57, (Rat) Q32Q05, (Mouse) Q8CB27, (Zebrafish) Q567B1
Entrez Gene ID: (Human) 55432, (Dog) 490268, (Bovine) 539931, (Rabbit) 100344429, (Rat) 363982, (Mouse) 226418, (Guinea pig) 100714320, (Zebrafish) 550411
Molecular Function:
hydrolase
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.