Invitrogen
Manufacturer Code:PA535042
Catalog # PIPA535042
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 319-347 amino acids from the C-terminal region of human YOD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DUBA-8; HIN-7; HIV-1-induced protease 7; hsHIN7; OTU domain containing 2; OTU domain-containing protein 2; Ubiquitin thioesterase OTU1; YOD1 DUBA8 HIN7 OTUD2 Ubiquitin thioesterase OTU1 DUBA-8 HIV-1-induced protease 7 OTU domain-containing protein 2; YOD1 OTU deubiquinating enzyme 1 homolog ( yeast)
Gene Aliases: DUBA8; HIN7; OTUD2; PRO0907; YOD1
UniProt ID: (Human) Q5VVQ6
Entrez Gene ID: (Human) 55432
Molecular Function: hydrolase protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.