Invitrogen
Manufacturer Code:PA5116139
Catalog # PIPA5116139
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human YOD1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9930028C20Rik DUBA8 DUBA-8 HIN7 HIN-7 HIV-1-induced protease 7 HIV-induced protein-7-like protease Hshin7 OTU domain containing 2 OTU domain-containing protein 2 OTUD2 PRO0907 Ubiquitin thioesterase OTU1 Yod1 YOD1 deubiquitinase YOD1 OTU deubiquinating enzyme 1 homolog YOD1 OTU deubiquinating enzyme 1 homolog ( yeast) YOD1 OTU deubiquitinating enzyme 1 homologue YOD1 OTU deubiquitinating enzyme 1 homologue (S. cerevisiae); DUBA-8; HIN-7; HIV-1-induced protease 7; hsHIN7; OTU domain containing 2; OTU domain-containing protein 2; Ubiquitin thioesterase OTU1; YOD1 OTU deubiquinating enzyme 1 homolog ( yeast)
Gene Aliases: DUBA8; HIN7; OTUD2; PRO0907; YOD1
UniProt ID: (Human) Q5VVQ6
Entrez Gene ID: (Human) 55432
Molecular Function:
hydrolase
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.