Novus Biologicals
Manufacturer Code:NBP222326
Catalog # NBP222326
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen this antibody was made to maps to a region between residue 1350 to 1400 of human YLP Motif Containing 1 using the numbering given in entry NP_062535.2 (GeneID 56252). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C14orf170 YLPM1 YLP motif containing 1 ZAP113 ZAP3; Nuclear protein ZAP3; protein phosphatase 1, regulatory subunit 169; YLP motif-containing protein 1; ZAP113
Gene Aliases: C14orf170; PPP1R169; YLPM1; ZAP113; ZAP3
UniProt ID: (Human) P49750
Entrez Gene ID: (Human) 56252
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.