Invitrogen
Manufacturer Code:PA5113258
Catalog # PIPA5113258
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Y+L amino acid transporter 2 protein (473-515AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: amino acid permease; amino acid permease Cationic amino acid transporter y+ system KIAA0245 LAT-2 LAT3 SLC7A6 solute carrier family 7 (amino acid transporter light chain y+L system) member 6 solute carrier family 7 (cationic amino acid transporter y+ system) member 6 Solute carrier family 7 member 6 y(+)L-type amino acid transporter 2 Y+L amino acid transporter 2 Y+LAT2 y+LAT-2; Cationic amino acid transporter, y+ system; SLC7A6; solute carrier family 7 (amino acid transporter light chain, y+L system), member 6; solute carrier family 7 (cationic amino acid transporter, y+ system), member 6; Solute carrier family 7 member 6; solute carriersolute carrier family 7 (cationic amino acid transporter, y+ system), member 6; y(+)L-type amino acid transporter 2; Y+L amino acid transporter 2; y+LAT-2; Y+LAT2
Gene Aliases: AI643885; KIAA0245; LAT-2; LAT3; SLC7A6; y+LAT-2
UniProt ID: (Human) Q92536, (Mouse) Q8BGK6
Entrez Gene ID: (Human) 9057, (Mouse) 330836
Molecular Function:
amino acid transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.