Invitrogen
Manufacturer Code:PA563901
Catalog # PIPA563901
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human YJEFN3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ApoA-I-binding protein 2; apolipoprotein A1 binding protein; apolipoprotein A1 binding protein hYjeF&lowbarN3 hYjeF&lowbarN3-19p13.11 yjeF N-terminal domain-containing protein 3 yjeF&lowbarN3; hYjeF_N3; hYjeF_N3-19p13.11; YjeF N-terminal domain-containing protein 3; YjeF_N3
Gene Aliases: AIBP2; YJEFN3
UniProt ID: (Human) A6XGL0
Entrez Gene ID: (Human) 374887
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.