Invitrogen
Manufacturer Code:PA542234
Catalog # PIPA542234
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human YIPF6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein YIPF6; Yip1 domain family, member 6; YIP1 family member 6
Gene Aliases: A430107J06Rik; FinGER6; YIPF6; zgc:86904
UniProt ID: (Human) Q96EC8, (Bovine) A6QLC6, (Mouse) Q8BR70, (Rat) Q4QQU5, (Zebrafish) Q6IQ85
Entrez Gene ID: (Human) 286451, (Bovine) 540347, (Rabbit) 100342581, (Horse) 100066275, (Dog) 491931, (Guinea pig) 100727349, (Mouse) 77929, (Rat) 363476, (Zebrafish) 415186
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.