Invitrogen
Manufacturer Code:PA567301
Catalog # PIPA567301
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human YIPF5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FinGER5 SB140 SMAP-5 SMAP5 YIP1A YIP1 family member 5 YPT-interacting protein 1 A five-pass transmembrane protein localizing in the Golgi apparatus and the endoplasmic reticulum 5 golgi membrane protein SB140 protein YIPF5 smooth muscle cell associated protein 5 smooth muscle cell-associ; Five-pass transmembrane protein localizing in the Golgi apparatus and the endoplasmic reticulum 5; golgi membrane protein SB140; Protein YIPF5; SMAP-5; smooth muscle cell associated protein 5; Smooth muscle cell-associated protein 5; Yip1 domain family, member 5; YIP1 family member 5; YPT-interacting protein 1 A
Gene Aliases: FINGER5; PP12723; SB140; SMAP-5; SMAP5; UNQ3123/PRO10275; YIP1A; YIPF5
UniProt ID: (Human) Q969M3
Entrez Gene ID: (Human) 81555
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.