Invitrogen
Manufacturer Code:PA5113212
Catalog # PIPA5113212
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Protein YIPF5 protein (1-124AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610311I19Rik AA408236 Ac2-256 FINGER5 Five-pass transmembrane protein localizing in the Golgi apparatus and the endoplasmic reticulum 5 golgi membrane protein SB140 PP12723 Protein YIPF5 SB140 SMAP5 SMAP-5 smooth muscle cell associated protein 5 Smooth muscle cell-associated protein 5 UNQ3123/PRO10275 Yip1 domain family member 5 Yip1 domain family member 5 YIP1 family member 5 YIP1A Yipf5 YPT-interacting protein 1 A; Five-pass transmembrane protein localizing in the Golgi apparatus and the endoplasmic reticulum 5; golgi membrane protein SB140; Protein YIPF5; SMAP-5; smooth muscle cell associated protein 5; Smooth muscle cell-associated protein 5; Yip1 domain family, member 5; YIP1 family member 5; YPT-interacting protein 1 A
Gene Aliases: 2610311I19Rik; AA408236; FINGER5; PP12723; SB140; SMAP-5; SMAP5; UNQ3123/PRO10275; YIP1A; YIPF5
UniProt ID: (Human) Q969M3, (Mouse) Q9EQQ2
Entrez Gene ID: (Human) 81555, (Mouse) 67180
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.