Proteintech
Manufacturer Code:154731AP150UL
Catalog # 50-173-0256
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | YIPF4 Fusion Protein Ag7777 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FinGER4 Nbla11189 Protein YIPF4 Yip1 domain family member 4 YIP1 family member 4 YIPF4; Protein YIPF4; putative protein product of Nbla11189; Yip1 domain family, member 4; YIP1 family member 4
Gene Aliases: 2310034L04Rik; FinGER4; Nbla11189; RGD1310157; YIPF4
UniProt ID: (Human) Q9BSR8, (Mouse) Q8C407, (Rat) Q5M7T4
Entrez Gene ID: (Human) 84272, (Mouse) 67864, (Rat) 362699
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.