Invitrogen
Manufacturer Code:MA537696
Catalog # PIMA537696
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1612CT505.7.50 |
Immunogen | recombinant protein of human YES1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cellular yes-1 protein; cellular yes-1 protein c-Src c-yes EC 2.7.10.2 HsT441 p60c-yes p60-Src p61-Yes pp60c-src Proto-oncogene c-Yes proto-oncogene tyrosine-protein kinase Yes tyrosine-protein kinase Yes viral oncogene yes homolog v-yes-1 Yamaguchi sarcoma viral oncogene homolog 1 Yamagichi sarcoma viral (v-yes-1) oncogene homolog 1 Yamaguchi sarcoma oncogene Yamaguchi sarcoma viral (v-yes) oncogene homolog 1 Yamaguchi sarcoma viral oncogene homolog 1 YES YES proto-oncogene 1 Src family tyrosine kinase YES1 YES1 proto-oncogene Src family tyrosine kinase; p61-Yes; Proto-oncogene c-Yes; proto-oncogene tyrosine-protein kinase YES; Tyrosine-protein kinase Yes; v-yes-1 Yamaguchi sarcoma viral oncogene homolog 1; Yamaguchi sarcoma oncogene; YES1 proto-oncogene, Src family tyrosine kinase
Gene Aliases: c-yes; HsT441; P61-YES; YES; YES1
UniProt ID: (Human) P07947
Entrez Gene ID: (Human) 7525
Molecular Function: kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.