Invitrogen
Manufacturer Code:PA530614
Catalog # PIPA530614
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 62 and 287 of Human YBX2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Contrin; CONTRIN CSDA3 DBPC MSY2 DNA-binding protein C MSY2 homolog Y-box-binding protein 2 germ cell specific Y-box binding protein germ cell-specific Y-box-binding protein; Dbpc; DNA-binding protein C; FRGY2 homolog; germ cell specific Y-box binding protein; Germ cell-specific Y-box-binding protein; MSY2 homolog; Y box binding protein 2; Y-box protein MSY2; Y-box-binding protein 2
Gene Aliases: CONTRIN; CSDA3; DBPC; MSY2; YBX2
UniProt ID: (Human) Q9Y2T7
Entrez Gene ID: (Human) 51087, (Mouse) 53422
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.