Novus Biologicals
Manufacturer Code:NBP298908B
Catalog # NB106669
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human YB1. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BP-8 CBF-A class II Y box-binding protein I CSDA2 CSDB DBPB CCAAT-binding transcription factor I subunit A EFI-A Enhancer factor I subunit A MDR-NF1 MGC104858 MGC110976 MGC117250 NSEP1 Y-box-binding protein 1 nuclease-sensitive element-binding protein 1 Y box binding protein 1 YB1 DNA-binding protein B YB-1 Y-box transcription factor YBX1; CBF-A; CCAAT-binding transcription factor I subunit A; DBPB; DNA-binding protein B; EFI-A; Enhancer factor I subunit A; major histocompatibility complex, class II, Y box-binding protein I; nuclease sensitive element binding protein 1; Nuclease-sensitive element-binding protein 1; Y box binding protein 1; Y-box transcription factor; Y-box-binding protein 1; YB-1
Gene Aliases: BP-8; CBF-A; CSDA2; CSDB; DBPB; EFI-A; MDR-NF1; NSEP-1; NSEP1; YB-1; YB1; YBX1
UniProt ID: (Human) P67809
Entrez Gene ID: (Human) 4904
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.