Abnova
Manufacturer Code:H00051067B01P
Catalog # 89-935-117
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | YARS2 (NP_001035526.11 a.a. &bsim 477 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human YARS2 protein.
For Research Use Only
Protein Aliases: CGI-04; FLJ13995; mt-TyrRS; tyrosine tRNA ligase 2, mitochondrial; Tyrosine--tRNA ligase, mitochondrial; Tyrosyl-tRNA synthetase; tyrosyl-tRNA synthetase 2, mitochondrial; TYRRS
Gene Aliases: CGI-04; MLASA2; MT-TYRRS; TYRRS; YARS2
UniProt ID: (Human) Q9Y2Z4
Entrez Gene ID: (Human) 51067
Molecular Function:
aminoacyl-tRNA synthetase
ligase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.