Proteintech
Manufacturer Code:171501AP150UL
Catalog # 50-173-1406
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | YARS2 Fusion Protein Ag9530 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CGI 04 mt TyrRS Tyrosine tRNA ligase TYRRS YARS2; tyrosine tRNA ligase 2, mitochondrial; Tyrosine--tRNA ligase, mitochondrial; Tyrosyl-tRNA synthetase; tyrosyl-tRNA synthetase 2, mitochondrial; tyrosyl-tRNA synthetase, mitochondrial; TyrRS
Gene Aliases: 2210023C10Rik; CGI-04; MLASA2; MT-TYRRS; RGD1311696; TYRRS; YARS2
UniProt ID: (Human) Q9Y2Z4, (Mouse) Q8BYL4, (Rat) Q5I0L3
Entrez Gene ID: (Human) 51067, (Mouse) 70120, (Rat) 287924
Molecular Function:
aminoacyl-tRNA synthetase
ligase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.