Invitrogen
Manufacturer Code:PA553882
Catalog # PIPA553882
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human YARS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CMTDIC TYRRS YRS YTS tyrosine tRNA ligase 1 cytoplasmic tyrosine--tRNA ligase cytoplasmic tyrosyl--tRNA ligase tyrosyl-tRNA synthetase cytoplasmic; tyrosine tRNA ligase 1, cytoplasmic; Tyrosine--tRNA ligase, cytoplasmic; Tyrosine--tRNA ligase, cytoplasmic, N-terminally processed; tyrosyl--tRNA ligase; Tyrosyl-tRNA synthetase; tyrosyl-tRNA synthetase, cytoplasmic; TyrRS
Gene Aliases: AL024047; AU018965; CMTDIC; TYRRS; YARS; YARS1; YRS; YTS
UniProt ID: (Human) P54577, (Mouse) Q91WQ3, (Rat) Q4KM49
Entrez Gene ID: (Human) 8565, (Mouse) 107271, (Rat) 313047
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.