R&D Systems
Manufacturer Code:MAB8094
Catalog # MAB8094
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 867711 |
Immunogen | <EM>E. coli</EM>-derived recombinant human YAP1 Ala347-Leu504 Accession # P46937 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 65 kDa Yes-associated protein; 65 kDa Yes-associated protein COB1 Protein Yorkie Homolog Transcriptional Coactivator YAP1 YAP YAP1-2gamma YAP2 YAP2L YAP65 YAP65yes-associated protein 2 Yes Associated Protein Yes-Associated Protein Yes-associated protein 1 Yes-associated protein 1 65kDa yes-associated protein beta yes-associated protein delta Yes-Associated Protein YAP65 Homolog YKI Yorkie Homolog; Protein yorkie homolog; Transcriptional coactivator YAP1; Yes-associated protein 1; yes-associated protein 2; Yes-associated protein YAP65 homolog; yorkie homolog
Gene Aliases: COB1; YAP; YAP1; YAP2; YAP65; YKI
UniProt ID: (Human) P46937
Entrez Gene ID: (Human) 10413
Molecular Function:
enzyme modulator
kinase modulator
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.