Invitrogen
Manufacturer Code:PA578322
Catalog # PIPA578322
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human YAP1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 65 kDa Yes-associated protein; Protein yorkie homolog; Transcriptional coactivator YAP1; yap; YAP65 YAP Yes-associated protein 1 YAP2 YKI; Yes-associated protein 1; yes-associated protein 2; Yes-associated protein YAP65 homolog; yorkie homolog
Gene Aliases: cb194; CH211-181P1.5-001; COB1; sb:cb194; si:ch211-181p1.5; si:dkey-3b8.3; wu:fc18c04; YAP; YAP1; YAP2; YAP65; YKI; zgc:158380; zYAP
UniProt ID: (Human) P46937, (Zebrafish) Q1L8J7
Entrez Gene ID: (Human) 10413, (Zebrafish) 561411
Molecular Function:
enzyme modulator
kinase modulator
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.