Novus Biologicals
Manufacturer Code:NBP146105
Catalog # NBP146105
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this YAP1 antibody maps to a synthectic peptide with the sequence of C-TYHSRDESTDS (NP_006097). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 65 kDa Yes-associated protein; 65 kDa Yes-associated protein COB1 Protein Yorkie Homolog Transcriptional Coactivator YAP1 YAP YAP1-2gamma YAP2 YAP2L YAP65 YAP65yes-associated protein 2 Yes Associated Protein Yes-Associated Protein Yes-associated protein 1 Yes-associated protein 1 65kDa yes-associated protein beta yes-associated protein delta Yes-Associated Protein YAP65 Homolog YKI yorkie homolog; Protein yorkie homolog; Transcriptional coactivator YAP1; Yes-associated protein 1; yes-associated protein 2; Yes-associated protein YAP65 homolog; yorkie homolog
Gene Aliases: COB1; YAP; YAP1; YAP2; YAP65; YKI
UniProt ID: (Human) P46937
Entrez Gene ID: (Human) 10413
Molecular Function: enzyme modulator kinase modulator transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.