Abnova
Manufacturer Code:PAB9591
Catalog # 89-134-256
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues surrounding amino acids 279 of human YAP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of YAP1.
For Research Use Only
Protein Aliases: 65 kDa Yes-associated protein; Protein yorkie homolog; Transcriptional coactivator YAP1; YAP; YAP2; YAP65; Yes-associated protein 1; yes-associated protein 2; Yes-associated protein YAP65 homolog; YKI; yorkie homolog
Gene Aliases: COB1; YAP; YAP1; YAP2; YAP65; YKI
UniProt ID: (Human) P46937
Entrez Gene ID: (Human) 10413
Molecular Function:
enzyme modulator
kinase modulator
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.