Novus Biologicals
Manufacturer Code:NBP297898AF532
Catalog # NB049137
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human Xylulokinase/XYLB fragment. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D-xylulokinase; EC 2.7.1.17 FLJ10343 FLJ12539 FLJ22075 Xylulokinase xylulokinase (H. influenzae) homolog xylulokinase homolog (H. influenzae) xylulose kinase; Xylulokinase; xylulokinase homolog (H. influenzae); Xylulose kinase
Gene Aliases: XYLB
UniProt ID: (Human) O75191
Entrez Gene ID: (Human) 9942
Molecular Function:
carbohydrate kinase
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.