Abnova
Manufacturer Code:PAB15694
Catalog # 89-117-759
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant GST fusion protein corresponding to 108 mouse Xpo5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant Xpo5.
For Research Use Only
Protein Aliases: 2410004H11Rik; 2700038C24Rik; AI648907; AW549301; Exp5; Exportin-5; mKIAA1291; Ran-binding protein 21; RanBp21
Gene Aliases: 2410004H11Rik; 2700038C24Rik; AI648907; AW549301; Exp5; Kiaa1291; mKIAA1291; RanBp21; Xpo5
UniProt ID: (Mouse) Q924C1
Entrez Gene ID: (Mouse) 72322
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.