Invitrogen
Manufacturer Code:PA543402
Catalog # PIPA543402
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human XYLT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Peptide O-xylosyltransferase 1; Peptide O-xylosyltransferase 2; protein xylosyltransferase 2; UDP-D-xylose:proteoglycan core protein beta-D-xylosyltransferase; XT-II; Xylosyltransferase 2; Xylosyltransferase II; Xylosyltransferase II XT-II XT2 xylT-II PXYLT2
Gene Aliases: E030002B02Rik; PXYLT2; SOS; UNQ3058/PRO9878; XT-II; XT2; xylT-II; XYLT2
UniProt ID: (Human) Q9H1B5, (Bovine) Q5QQ49, (Dog) Q5QQ50, (Mouse) Q9EPL0
Entrez Gene ID: (Human) 64132, (Bovine) 493989, (Rabbit) 100342073, (Pig) 100514793, (Horse) 100069884, (Dog) 493990, (Guinea pig) 100714934, (Rat) 64134, (Mouse) 217119
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.