Invitrogen
Manufacturer Code:PA529127
Catalog # PIPA529127
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 272 and 520 of Human Xylosyltransferase II |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Peptide O-xylosyltransferase 1; protein xylosyltransferase 2; PXYLT2 XT-II XT2 xylT-II UDP-D-xylose:proteoglycan core protein beta-D-xylosyltransferase peptide O-xylosyltransferase 1 protein xylosyltransferase 2 xylosyltransferase 2; UDP-D-xylose:proteoglycan core protein beta-D-xylosyltransferase; XT-II; Xylosyltransferase 2; Xylosyltransferase II
Gene Aliases: PXYLT2; SOS; UNQ3058/PRO9878; XT-II; XT2; xylT-II; XYLT2
UniProt ID: (Human) Q9H1B5
Entrez Gene ID: (Human) 64132
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.