Invitrogen
Manufacturer Code:PA567627
Catalog # PIPA567627
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human XYLT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: beta-D-xylosyltransferase 1; Peptide O-xylosyltransferase 1; XT-I; Xylosyltransferase 1; Xylosyltransferase 1 Peptide O-xylosyltransferase 1 Xylosyltransferase I XT-I XylT-I XYLT1 XT1; Xylosyltransferase I; xylosyltransferase iota
Gene Aliases: 8030490L12; DBQD2; PXYLT1; XT-I; XT1; XTI; XYLT-1; xylT-I; XYLT1; XYLTI
UniProt ID: (Human) Q86Y38, (Mouse) Q811B1
Entrez Gene ID: (Human) 64131, (Rat) 64133, (Mouse) 233781
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.