Invitrogen
Manufacturer Code:PA542855
Catalog # PIPA542855
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human XTP3TPA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dCTP pyrophosphatase 1; dCTPase 1; Deoxycytidine-triphosphatase 1; RS21C6; XTP3-transactivated gene A protein; XTP3-transactivated gene A protein homolog; XTP3-transactivated protein A
Gene Aliases: CDA03; DCTPP1; RS21-C6; RS21C6; XTP3TPA
UniProt ID: (Human) Q9H773, (Bovine) Q32KY6
Entrez Gene ID: (Human) 79077, (Horse) 100065556, (Dog) 489903, (Bovine) 614103, (Guinea pig) 100729302
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.