Novus Biologicals
Manufacturer Code:NBP276375
Catalog # NBP276375
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Immunogen sequence between amino acids 120 abd 170 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 2-8°C/ 1 year from date of receipt |
Rabbit Polyclonal Antibody
Protein Aliases: dCTP pyrophosphatase 1; dCTP pyrophosphatase 1 dCTPase 1 Deoxycytidine-triphosphatase 1 EC 3.6.1.12 MGC5627 RS21C6XTP3-transactivated gene A protein XTP3TPACDA03 XTP3-transactivated protein A; dCTPase 1; Deoxycytidine-triphosphatase 1; RS21C6; XTP3-transactivated gene A protein; XTP3-transactivated protein A
Gene Aliases: CDA03; DCTPP1; RS21C6; XTP3TPA
UniProt ID: (Human) Q9H773
Entrez Gene ID: (Human) 79077
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.