Abnova
Manufacturer Code:H00079077B01P
Catalog # 89-107-315
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | XTP3TPA (NP_077001.11 a.a. &bsim 170 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human XTP3TPA protein.
For Research Use Only
Protein Aliases: CDA03; dCTP pyrophosphatase 1; dCTPase 1; Deoxycytidine-triphosphatase 1; MGC5627; RS21C6; XTP3-transactivated gene A protein; XTP3-transactivated protein A; XTP3TPA
Gene Aliases: CDA03; DCTPP1; RS21C6; XTP3TPA
UniProt ID: (Human) Q9H773
Entrez Gene ID: (Human) 79077
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.