Invitrogen
Manufacturer Code:PA569958
Catalog # PIPA569958
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of human XRRA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: X-ray radiation resistance associated 1 protein; X-ray radiation resistance-associated protein 1; XRRA1
Gene Aliases: XRRA1
UniProt ID: (Human) Q6P2D8, (Bovine) Q7PCK7
Entrez Gene ID: (Rabbit) 100355342, (Human) 143570, (Bovine) 369019, (Horse) 100064954, (Dog) 476805, (Rat) 365313, (Guinea pig) 100734032
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.