Invitrogen
Manufacturer Code:MA527148
Catalog # PIMA527148
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5F4 |
Immunogen | Human recombit protein fragment corresponding to amino acids 677-779 of XRN2 produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 5'-3' exoribonuclease 2; DHM1-like protein; DHP protein; Protein Dhm1; XRN2b
Gene Aliases: Dhm1; XRN2
UniProt ID: (Human) Q9H0D6, (Mouse) Q9DBR1
Entrez Gene ID: (Human) 22803, (Mouse) 24128
Molecular Function: RNA binding protein exoribonuclease nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.