Invitrogen
Manufacturer Code:PA5104231
Catalog # PIPA5104231
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human XRN2 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5'-3' exoribonuclease 2; 5'-3' exoribonuclease 2 Dhm1 DHM1-like protein DHP protein Protein Dhm1 Xrn2 XRN2b; DHM1-like protein; DHP protein; Protein Dhm1
Gene Aliases: Dhm1; XRN2
UniProt ID: (Human) Q9H0D6, (Mouse) Q9DBR1
Entrez Gene ID: (Human) 22803, (Mouse) 24128
Molecular Function:
RNA binding protein
exoribonuclease
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.