Novus Biologicals
Manufacturer Code:NB10057539
Catalog # NB10057539
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human 5'-3' exoribonuclease 2 using the numbering given in entry NP_036387.2 (GeneID 22803). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5'-3' exoribonuclease 2; 5'-3' exoribonuclease 2 DHM1-like protein Dhm1-like protein (mouse homolog) DHP protein EC 3.1.13 EC 3.1.13.-; DHM1-like protein; DHP protein
Gene Aliases: XRN2
UniProt ID: (Human) Q9H0D6
Entrez Gene ID: (Human) 22803
Molecular Function:
RNA binding protein
exoribonuclease
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.