Invitrogen
Manufacturer Code:PA576068
Catalog # PIPA576068
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human XRCC4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA repair protein XRCC4; DNA repair protein XRCC4 X-ray repair cross-complementing protein 4 XRCC4; X-ray repair complementing defective repair in Chinese hamster cells 4; X-ray repair cross complementing protein 4; X-ray repair cross-complementing protein 4
Gene Aliases: 2310057B22Rik; AW413319; AW545101; SSMED; XRCC4
UniProt ID: (Human) Q13426, (Mouse) Q924T3
Entrez Gene ID: (Human) 7518, (Mouse) 108138, (Rat) 309995
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.