OriGene
Manufacturer Code:TA804763
Catalog # 50-168-0392
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5B11 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-235 of human XRCC3 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CMM6; DNA repair protein XRCC3; X-ray repair complementing defective repair in Chinese hamster cells 3; X-ray repair cross-complementing protein 3
Gene Aliases: CMM6; XRCC3
UniProt ID: (Human) O43542
Entrez Gene ID: (Human) 7517
Molecular Function: DNA binding protein DNA strand-pairing protein hydrolase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.