Invitrogen
Manufacturer Code:PA585194
Catalog # PIPA585194
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human XRCC3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA repair protein XRCC3; X-ray repair complementing defective repair in Chinese hamster cells 3; X-ray repair complementing defective repair in Chinese hamster cells 3 Antibody CMM6 Antibody; X-ray repair cross-complementing protein 3
Gene Aliases: CMM6; XRCC3
UniProt ID: (Human) O43542
Entrez Gene ID: (Human) 7517
Molecular Function:
DNA binding protein
DNA strand-pairing protein
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.