Invitrogen
Manufacturer Code:PA575934
Catalog # PIPA575934
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 127-172 of Human XRCC3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA repair protein XRCC3; DNA repair protein XRCC3 X-ray repair cross-complementing protein 3 XRCC3; DNA repair protein XRCC3-like; X-ray repair complementing defective repair in Chinese hamster cells 3; X-ray repair cross-complementing protein 3
Gene Aliases: 4432412E01Rik; AI182522; AW537713; CMM6; XRCC3
UniProt ID: (Human) O43542, (Mouse) Q9CXE6
Entrez Gene ID: (Rat) 102551085, (Human) 7517, (Mouse) 74335
Molecular Function:
DNA binding protein
DNA strand-pairing protein
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.