Novus Biologicals
Manufacturer Code:NB100165SS
Catalog # NB100165SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic partial peptide of human XRCC3. [UniProt# O43542] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CMM6 DNA repair protein XRCC3 X-ray repair complementing defective repair in Chinese hamster cells 3 X-ray repair cross-complementing protein 3; DNA repair protein XRCC3; X-ray repair complementing defective repair in Chinese hamster cells 3; X-ray repair cross-complementing protein 3
Gene Aliases: CMM6; XRCC3
UniProt ID: (Human) O43542
Entrez Gene ID: (Human) 7517
Molecular Function:
DNA binding protein
DNA strand-pairing protein
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.