Invitrogen
Manufacturer Code:PA576067
Catalog # PIPA576067
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human XRCC2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA repair protein XRCC2; DNA repair protein XRCC2 X-ray repair cross-complementing protein 2 XRCC2; recA/RAD51 family protein; X-ray repair complementing defective repair in Chinese hamster cells 2; X-ray repair cross complementing protein 2; X-ray repair cross-complementing protein 2
Gene Aliases: 4921524O04Rik; 8030409M04Rik; FANCU; RAD51; RecA; RGD1564823; XRCC2
UniProt ID: (Human) O43543, (Mouse) Q9CX47
Entrez Gene ID: (Human) 7516, (Mouse) 57434, (Rat) 499966
Molecular Function: DNA binding protein DNA strand-pairing protein hydrolase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.