Novus Biologicals
Manufacturer Code:NBP256170
Catalog # NBP256170
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:AQLVNGVGGGKVESLLRRAFGAMCSAFHRAESGTELLARLEGRSSLKEIEPNLFADEDSPVHGDILEFHGPEGTGKTEMLYHLTARCILPKSEGGLEVEVLFIDTDYHFDMLRLVTILEHRLSQSSEEIIKYC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp781P0919 DNA repair protein XRCC2 X-ray repair complementing defective repair in Chinese hamster cells 2 X-ray repair cross-complementing protein 2 X-ray repair complementing defective repair in Chinese hamster; DNA repair protein XRCC2; X-ray repair complementing defective repair in Chinese hamster cells 2; X-ray repair cross-complementing protein 2
Gene Aliases: FANCU; XRCC2
UniProt ID: (Human) O43543
Entrez Gene ID: (Human) 7516
Molecular Function: DNA binding protein DNA strand-pairing protein hydrolase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.