Invitrogen
Manufacturer Code:PA580237
Catalog # PIPA580237
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human XRCC1 recombit protein (Position: E538-A633). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA repair protein XRCC1; DNA repair protein XRCC1 X-ray repair cross-complementing protein 1 XRCC1; X-ray repair complementing defective repair in Chinese hamster cells 1; x-ray repair cross-complementing group 1 protein; X-ray repair cross-complementing protein 1
Gene Aliases: RCC; Xrcc-1; XRCC1
UniProt ID: (Human) P18887, (Rat) Q9ESZ0, (Mouse) Q60596
Entrez Gene ID: (Human) 7515, (Rat) 84495, (Mouse) 22594
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.