Invitrogen
Manufacturer Code:PA525458
Catalog # PIPA525458
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 407-435 amino acids from the central region of human XRCC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA repair protein XRCC1; RCC DNA repair protein XRCC1 X-ray repair cross-complementing protein 1 X-ray-repair complementing defective repair in Chinese hamster; X-ray repair complementing defective repair in Chinese hamster cells 1; X-ray repair cross-complementing protein 1
Gene Aliases: RCC; XRCC1
UniProt ID: (Human) P18887
Entrez Gene ID: (Human) 7515
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.