Novus Biologicals
Manufacturer Code:NBP29400120UL
Catalog # NB022568
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 464-631 of human XPR1 (NP_001129141.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ90308 Protein SYG1 homolog SYG1xenotropic and polytropic retrovirus receptor X3 Xenotropic and polytropic murine leukemia virus receptor X3 xenotropic and polytropic retrovirus receptor 1 XR X-receptor; Protein SYG1 homolog; X-receptor; Xenotropic and polytropic murine leukemia virus receptor X3; Xenotropic and polytropic retrovirus receptor 1
Gene Aliases: IBGC6; SYG1; X3; XPR1; XR
UniProt ID: (Human) Q9UBH6
Entrez Gene ID: (Human) 9213
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.